ENA100801081 Enamine

5-piperidin-1-yl-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691153
  • Cas Number : 923124-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25950

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25950
Availability details for
Quantity
  • Availability
  • Add to Cart