ENA100801094 Enamine

4-cyclopropyl-5-(4-methylpiperidin-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691177
  • Cas Number : 923121-82-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26011

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26011
Availability details for
Quantity
  • Availability
  • Add to Cart