ENA100801097 Enamine

5-pyrrolidin-1-yl-4-(tetrahydrofuran-2-ylmethyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691181
  • Cas Number : 923178-04-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26019

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26019
Availability details for
Quantity
  • Availability
  • Add to Cart