ENA100801100 Enamine

1'-cyclopentyl-6'-oxo-1',5',6',7'-tetrahydrospiro[cyclohexane-1,4'-pyrazolo[3,4-b]pyridine]-5'-carbonitrile

  • MDL number MFCD08691187
  • Cas Number : 923152-34-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26028

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26028
Availability details for
Quantity
  • Availability
  • Add to Cart