ENA100801101 Enamine

3-(phenoxymethyl)-2-furoic acid

  • MDL number MFCD08691188
  • Cas Number : 923129-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 129...131
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26029

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26029
Availability details for
Quantity
  • Availability
  • Add to Cart