ENA100801106 Enamine

[6-(1H-pyrazol-1-yl)pyridin-3-yl]methanamine

  • MDL number MFCD08691193
  • Cas Number : 956191-88-1
  • Molecular Weight : 174.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-26039

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26039
Availability details for
Quantity
  • Availability
  • Add to Cart