ENA100801124 Enamine

4-(2-furylmethyl)-5-(3-methylpiperidin-1-yl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691213
  • Cas Number : 923805-80-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-26119

Safety Information

msds https://www.enaminestore.com/catalog/EN300-26119
Availability details for
Quantity
  • Availability
  • Add to Cart