ENA100801125 Enamine

2-chloro-N-(3-cyano-1-cyclopentyl-4,5-dimethyl-1H-pyrrol-2-yl)acetamide

  • MDL number MFCD08691214
  • Cas Number : 923190-03-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-26120

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-26120
Availability details for
Quantity
  • Availability
  • Add to Cart