ENA100801434 Enamine

1,3,5-trimethyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD00159648
  • Cas Number : 1125-29-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25595

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25595
Availability details for
Quantity
  • Availability
  • Add to Cart