ENA100801437 Enamine

2-chloro-1-[4-(3-chloro-4-fluorobenzenesulfonyl)piperazin-1-yl]ethan-1-one

  • MDL number MFCD08691087
  • Cas Number : 923780-46-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-22973

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-22973
Availability details for
Quantity
  • Availability
  • Add to Cart