ENA100801459 Enamine

2-({4-[(dimethylamino)sulfonyl]benzoyl}amino)-4-methylpentanoic acid

  • MDL number MFCD08444431
  • Cas Number : 1009540-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25608

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25608
Availability details for
Quantity
  • Availability
  • Add to Cart