ENA100801460 Enamine

2-{[2-chloro-5-(dimethylsulfamoyl)phenyl]formamido}propanoic acid

  • MDL number MFCD08444432
  • Cas Number : 1008675-36-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-25609

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25609
Availability details for
Quantity
  • Availability
  • Add to Cart