ENA100801469 Enamine

2-cyclopropylquinazoline-4-thiol

  • MDL number MFCD03825835
  • Cas Number : 604754-96-3
  • Molecular Weight : 202.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-25614

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25614
Availability details for
Quantity
  • Availability
  • Add to Cart