ENA100801485 Enamine

methyl 2-[(chloroacetyl)amino]-3-methylpentanoate

  • MDL number MFCD00548262

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23149

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23149
Availability details for
Quantity
  • Availability
  • Add to Cart