ENA100801487 Enamine

2-chloro-N-[2-(dimethylamino)-5-(piperidine-1-sulfonyl)phenyl]acetamide

  • MDL number MFCD08444136
  • Cas Number : 923805-28-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23159

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23159
Availability details for
Quantity
  • Availability
  • Add to Cart