ENA100801488 Enamine

2-Morpholin-4-yl-5-trifluoromethyl-phenylamine

  • MDL number MFCD00042160
  • Cas Number : 784-57-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 133...135
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-02435

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-02435
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart