ENA100801494 Enamine

{[5-(4-chlorophenyl)-4-(2-furylmethyl)-4H-1,2,4-triazol-3-yl]thio}acetic acid

  • MDL number MFCD08444445
  • Cas Number : 537017-56-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25641

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25641
Availability details for
Quantity
  • Availability
  • Add to Cart