ENA100801496 Enamine

[(4-isopropyl-4H-1,2,4-triazol-3-yl)thio]acetic acid

  • MDL number MFCD08444446
  • Cas Number : 923232-99-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25643

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25643
Availability details for
Quantity
  • Availability
  • Add to Cart