ENA100801500 Enamine

1-(4-fluorophenyl)-1H-pyrazolo[3,4-d]pyrimidine-4-thiol

  • MDL number MFCD08691117
  • Cas Number : 1955554-42-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25646

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25646
Availability details for
Quantity
  • Availability
  • Add to Cart