ENA100801504 Enamine

3-(4-chlorophenyl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD07367328
  • Cas Number : 135641-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25651

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25651
Availability details for
Quantity
  • Availability
  • Add to Cart