ENA100801507 Enamine

2-chloro-N-(2-methanesulfonylphenyl)acetamide

  • MDL number MFCD08444147
  • Cas Number : 923175-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23249

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23249
Availability details for
Quantity
  • Availability
  • Add to Cart