ENA100801510 Enamine

2-chloro-N-{2-methyl-1-[4-(propan-2-yl)phenyl]propyl}acetamide

  • MDL number MFCD08691099
  • Cas Number : 923248-45-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23281

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23281
Availability details for
Quantity
  • Availability
  • Add to Cart