ENA100801512 Enamine

6-(4-fluorophenyl)-3-methylisoxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08444455
  • Cas Number : 923909-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25661
Availability details for
Quantity
  • Availability
  • Add to Cart