ENA100801514 Enamine

6-(2-fluorophenyl)-3-methylisoxazolo[5,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08444456
  • Cas Number : 924129-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25662

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25662
Availability details for
Quantity
  • Availability
  • Add to Cart