ENA100801543 Enamine

4-(2,3-dihydro-1-benzofuran-5-yl)-1,3-thiazol-2-amine

  • MDL number MFCD03424576
  • Cas Number : 436151-87-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 173...175
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25679

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25679
Availability details for
Quantity
  • Availability
  • Add to Cart