ENA100801545 Enamine

methyl 2-[(chloroacetyl)amino]-3-(1H-indol-3-yl)propanoate

  • MDL number MFCD01566900
  • Cas Number : 108273-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 130...132
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-23443
Availability details for
Quantity
  • Availability
  • Add to Cart