ENA100801547 Enamine

2-chloro-N-[2-(2,3-dihydro-1,4-benzodioxin-6-yl)ethyl]acetamide

  • MDL number MFCD08444178
  • Cas Number : 916203-05-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 82-83
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23455

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23455
Availability details for
Quantity
  • Availability
  • Add to Cart