ENA100801551 Enamine

4-(2-chloroacetyl)-N-phenylpiperazine-1-carboxamide

  • MDL number MFCD08444180
  • Cas Number : 923125-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-23471

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-23471
Availability details for
Quantity
  • Availability
  • Add to Cart