ENA100801569 Enamine

5-(1,2,3,4-tetrahydroisoquinoline-2-sulfonyl)thiophene-3-carboxylic acid

  • MDL number MFCD08444479
  • Cas Number : 924062-42-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25707

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25707
Availability details for
Quantity
  • Availability
  • Add to Cart