ENA100801571 Enamine

[(1-cyclopentyl-1H-tetrazol-5-yl)thio]acetic acid

  • MDL number MFCD03566949
  • Cas Number : 531536-55-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25711

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25711
Availability details for
Quantity
  • Availability
  • Add to Cart