ENA100801590 Enamine

1-(2-methyl-1,3-thiazol-4-yl)ethan-1-one

  • MDL number MFCD00278580
  • Cas Number : 23002-78-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 55-57
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25717

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25717
Availability details for
Quantity
  • Availability
  • Add to Cart