ENA100801630 Enamine

2-bromo-1-cyclohexylethanone

  • MDL number MFCD08444491
  • Cas Number : 56077-28-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25739

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-25739
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart