ENA100801639 Enamine

2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]acetamide

  • MDL number MFCD01814799
  • Cas Number : 477888-16-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25743
Availability details for
Quantity
  • Availability
  • Add to Cart