ENA100801668 Enamine

N-(7-bromo-2,3-dihydro-1,4-benzodioxin-6-yl)-2-chloroacetamide

  • MDL number MFCD03004701
  • Cas Number : 923205-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24575
Availability details for
Quantity
  • Availability
  • Add to Cart