ENA100801679 Enamine

N,N-diethyl-2-hydroxy-5-nitrobenzene-1-sulfonamide

  • MDL number MFCD07696840
  • Cas Number : 180606-71-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25777
Availability details for
Quantity
  • Availability
  • Add to Cart