ENA100801726 Enamine

2-chloro-N-(2-oxo-2,3-dihydro-1H-1,3-benzodiazol-5-yl)acetamide

  • MDL number MFCD00823663
  • Cas Number : 339293-46-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230-231
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-08076

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-08076
Availability details for
Quantity
  • Availability
  • Add to Cart