ENA100801734 Enamine

N-phenyl-N'-(pyridin-3-ylmethyl)thiourea

  • MDL number MFCD00088576
  • Cas Number : 201215-93-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25455

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25455
Availability details for
Quantity
  • Availability
  • Add to Cart