ENA100801775 Enamine

1-isopropyl-6-(2-methylphenyl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08691148
  • Cas Number : 924243-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25933

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25933
Availability details for
Quantity
  • Availability
  • Add to Cart