ENA100801777 Enamine

2-{5H,6H,7H,8H,9H-[1,2,4]triazolo[4,3-a]azepin-3-yl}ethan-1-amine

  • MDL number MFCD08444080
  • Cas Number : 923255-73-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 63-64
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-25935

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25935
Availability details for
Quantity
  • Availability
  • Add to Cart