ENA100801779 Enamine

(1E)-2-(1H-imidazol-2-yl)-1-phenylethanone oxime

  • MDL number MFCD06378237
  • Cas Number : 851288-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25938

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25938
Availability details for
Quantity
  • Availability
  • Add to Cart