ENA100801803 Enamine

4-cyclopropyl-5-morpholin-4-yl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691155
  • Cas Number : 923237-40-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25953

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25953
Availability details for
Quantity
  • Availability
  • Add to Cart