ENA100801806 Enamine

2,2,2-trifluoroethyl 4-methylphenylcarbamate

  • MDL number MFCD07692334
  • Cas Number : 537-83-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 90...92
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-16350

Safety Information

msds https://www.enaminestore.com/catalog/EN300-16350
Availability details for
Quantity
  • Availability
  • Add to Cart