ENA100801807 Enamine

4-cyclopropyl-5-pyrrolidin-1-yl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691158
  • Cas Number : 923215-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25956

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25956
Availability details for
Quantity
  • Availability
  • Add to Cart