ENA100801808 Enamine

(1Z)-2-anilino-N,N'-diphenylethanimidamide

  • MDL number MFCD01110225
  • Cas Number : 14181-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25959

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25959
Availability details for
Quantity
  • Availability
  • Add to Cart