ENA100801825 Enamine

2-[(methylamino)methyl]thieno[3,2-d]pyrimidin-4(3H)-one

  • MDL number MFCD08691160
  • Cas Number : 923216-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25974

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25974
Availability details for
Quantity
  • Availability
  • Add to Cart