ENA100801826 Enamine

N1-[2-(dimethylamino)ethyl]benzene-1,2-diamine

  • MDL number MFCD08691161
  • Cas Number : 78156-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25975

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25975
Availability details for
Quantity
  • Availability
  • Add to Cart