ENA100801831 Enamine

2-chloro-5-(2,5-dimethyl-1H-pyrrol-1-yl)benzoic acid

  • MDL number MFCD01125265
  • Cas Number : 247225-32-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 140-142
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25540

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25540
Availability details for
Quantity
  • Availability
  • Add to Cart