ENA100801838 Enamine

1-ethyl-6-(thiophen-2-yl)-1H-pyrazolo[3,4-b]pyridine-4-carboxylic acid

  • MDL number MFCD08691168
  • Cas Number : 923919-56-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25987

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25987
Availability details for
Quantity
  • Availability
  • Add to Cart