ENA100801870 Enamine

N-tert-butyl-6-chloropyridine-3-sulfonamide

  • MDL number MFCD06292983
  • Cas Number : 622797-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-25550

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25550
Availability details for
Quantity
  • Availability
  • Add to Cart