ENA100801886 Enamine

2,2,2-trifluoroethyl N-(4-sulfamoylphenyl)carbamate

  • MDL number MFCD07685908
  • Cas Number : 872103-76-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 150-151
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-21777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-21777
Availability details for
Quantity
  • Availability
  • Add to Cart